Would you like to log in again?
Disposable Paper Funnels, Chemglass
Supplier: Chemglass
CLS-2080-100
10034-196PK 37.77 USD InStock
10034-196
Disposable Paper Funnels, Chemglass
Funnels Powder Funnels
The innovative 'backrest' of these sturdy, paper funnels allows hands-free use when pouring viscous liquids. With a 0.25" opening at the bottom, they fit a variety of vessels but are easily cut to any desired opening size or shape.
  • Made from clay-coated, heavy paper stock
  • Use with virtually any liquid or powder
  • Helps reduce cross contamination
  • Good alternative to disposable plastic funnels
  • Customizable spout can be trimmed to desired opening
  • Environmentally friendly; naturally biodegradable
Funnels stay in position with the application of a minimum amount of pressure. Use with virtually any liquid or powder, but they are not recommended for hot liquids. Supplied flat to maintain cleanliness and facilitate storage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty