Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NOS2 / iNOS (Human) ELISA Kit, BioVision

Supplier: BioVision
AB285315-96TEST
76305-640EA 1371.3 USD InStock
76305-640
NOS2 / iNOS (Human) ELISA Kit, BioVision
Assays ELISAs
A sandwich ELISA kit for the quantitative measurement of NOS2 in biological fluids.

  • Easy, convenient and time-saving method
  • Detection Range: 78.125 TO 5000 pg/ml
  • Sensitivity: <46.875 pg/ml
  • Assay precision: Intra-assay: CV <8%, inter-assay: CV <10%
  • No significant cross-reactivity or interference was observed

Nitric oxide synthases (NOS) are a family of enzymes that catalyze the production of nitric oxide (NO) from L-arginine. Nitric oxide (NO) plays an important role in neurotransmission, vascular regulation, immune response and apoptosis.

In presence of NADPH, FAD, FMN, (6R)-5,6,7,8-tetrahydrobiopterin, calmodulin and heme, NOS catalyzes a five-electron oxidation of the guanidino nitrogen of L-arginine with molecular oxygen to generate NO and L-citrulline. There are three isoforms of NOS: Endothelial (eNOS), neuronal (nNOS), and inducible (iNOS).

iNOS is expressed de novo under oxidative stress conditions and produces large amounts of NO as a part of body’s defense mechanism. NOS2 ELISA kit is a sandwich ELISA assay for the quantitative measurement of NOS2 in Human serum, plasma and cell culture supernatants.

The density of color is proportional to the amount of NOS2 captured from the samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty