Would you like to log in again?
Neomycin ELISA Kit, BioVision, Inc.
Supplier: BioVision
AB285276-96TEST
76305-662EA 1800.31 USD InStock
76305-662
Neomycin ELISA Kit, BioVision, Inc.
Assays ELISAs
A competitive ELISA kit for quantitate of neomycin in tissue, honey, milk, milk powder, liver.
  • Easy, convenient and time-saving method
  • Detection Range: 0.1 to 8.1 ppb
  • Sensitivity: <0.1 ppb
  • Detection limitation: 4 ppb
Neomycin is an aminoglycoside antibiotic found in many topical medications such as creams, ointments, and eyedrops. It is typically used as a topical preparation, such as neosporin. It can also be given orally, where it is usually combined with other antibiotics.
Neomycin, like other aminoglycosides, has been shown to be ototoxic causing tinnitus, hearing loss, and vestibular problems in a small number of patients. Standard techniques/instruments (HPLC or GC-MS) are utilized to detect neomycin.
However, these techniques are complex, expensive, laborious, and time-consuming. Immunoassay techniques, such as ELISAs are commonly preferred as a simple, reliable and rapid method for the quantification of neomycin in various samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty