Would you like to log in again?
AlphaTec® 37-200 General purpose gloves, Ansell
Supplier: Ansell
AlphaTec®
111701 135178 135181 135182 135179 135183 135180
76169-847CS 376.32 USD InStock
76169-847 76169-845 76169-839 76169-849 CA75877-330 76169-843 76169-841
AlphaTec® 37-200 General purpose gloves, Ansell
Gloves
Light weight gloves designed specifically for food processing and handling. Outstanding flexibility and dexterity, even at low temperatures.
  • Poses no risk to type one allergies
  • Unflocked lining to prevent the risk of food contamination
  • Liquid proof, can be used independently or as an outer glove
The innovative raised diamond surfacing of the UltraGrip™ technology offers an unmatched grip in wet and oily conditions. Ideal applications include diary processing, produce processing, poultry processing, and prepared meals.
Certifications: CE 0493 marked. ANSI Abrasion 4. Compliant with FDA food handling requirements.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty