Would you like to log in again?
AlphaTec® 87-297 Natural Rubber Latex Gloves with Cotton Lining, Yellow, Ansell
Supplier: Ansell
AlphaTec®
120191 120194 29711070 0029711080 29711100 120193 29711090 120192
76169-859CS 169.92 USD InStock
CA32890-542 CA32890-536 76169-859 CA32890-538 76169-861 76169-865 CA32890-540 76169-863
AlphaTec® 87-297 Natural Rubber Latex Gloves with Cotton Lining, Yellow, Ansell
Gloves
Gloves combining strength, sensitivity and grip. Medium duty, is both flexible and comfortable.
  • Material provides a high degree of tensile strength
  • 100% cotton flock lining helps maintain excellent sensitivity
  • Provides snag, puncture and abrasion resistance
  • Embossed pebble and fish scale designs provide improved grip on wet and dry objects
Ideal applications include chemical handling, clean room cleaning and preparing, and re-fuel oil and diesel.
Certifications: ANSI Abrasion 3. EN ISO 374-1 Type C. EN 388:2016 - 1010X. EN 388:2003 - 2110.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty