Would you like to log in again?
Marprene Tubing, Watson-Marlow
Supplier: Watson-Marlow
902.0064.016
MSPP-902064016BX 438.46 USD InStock
MSPP-902064016
Marprene Tubing, Watson-Marlow
Tubing Peristaltic Pump Tubing
Marprene tubing is ideal for high chemical resistance to acids and alkalis.
  • Wide chemical compatibility
  • Low gas permeability
  • Opaque to UV and visible light
It offers exceptionally long-life and resistance to a wide range of chemicals, especially oxidizing agents such as ozone, peroxides and sodium hypochlorite.
Operating pressures of 0 to 29 psi can be achieved with Marprene continuous tubing and has an operating temperature of +5 to +80 °C.
Certifications: Meets FDA requirement 21 CFR177.2600 for handling aqueous food and NSF/ANSI 61.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty