Would you like to log in again?
Whatman™ Protein Prep Syringe Filters for ӒKTA™ Systems, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10463103 10463033 10463043 10463033 10463113 10463043
76291-878PK 477.85 USD InStock
CA76291-876 76291-878 76291-876 CA76291-818 76291-880 76291-818
Whatman™ Protein Prep Syringe Filters for ӒKTA™ Systems, Whatman products (Cytiva)
Filters Syringe Filters
Protein prep syringe filters for ÄKTA™ protein purification systems are designed for optimal preparation of protein samples prior to chromatography.
  • Hydrophilic membrane made of regenerated cellulose exhibits low protein binding for sample preservation
  • Lot-to-lot certified to have low levels of extractables that could otherwise interfere with work
  • Filtration of samples prior to runs demonstrated to reduce column degrading particulate matter
High yield protein purification and accurate protein analysis both depend on a quality sample being introduced to the chromatography column. The protein prep syringe filter has been designed specifically to optimize these outcomes.
Ready-to-use with polycarbonate housing and a regenerated cellulose membrane that is low protein binding and broadly compatible with common solvents. Syringe filtration has been shown to reduce debris residue in the column that could otherwise impact performance and column life.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty