Would you like to log in again?
CREB (Phospho-Ser133) Transcription Factor Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical
10009846-96
75816-744EA 664.94 USD InStock
75816-744
CREB (Phospho-Ser133) Transcription Factor Assay Kit, Cayman Chemical Company
Assays ELISAs
A 96-well assay for measurement of CREB.
  • A sensitive, non-radioactive method of detecting CREB from whole cell lysates
  • 96-well plate format replaces EMSAs
  • Capture the transcription factor using a specific dsDNA sequence bound to the plate
  • Detect the dsDNA-bound transcription factor with specific antibodies in an ELISA format
  • Nuclear extraction kit is available to aid in the isolation of nuclear and cytoplasmic fractions from cell lysates or tissue homogenates
CREB (cAMP-response-element-binding protein) is a transcription factor that binds to cAMP-responsive element (CRE) promoter sites to regulate the transcription of numerous genes involved in metabolic regulation, depression, long term memory, and other physiological processes. Phosphorylation on serine 133 (Ser133) activates CREB to induce transcription of target genes. Diverse stimuli such as growth factors, neurotransmitters, hypoxia, growth factors, UV light, survival signals, and stress signals are some of the known activators of CREB. The signaling systems and mechanisms involved in CREB activation continue to be of interest in academic research and drug discovery programs. Cayman’s CREB (Phospho-Ser133) Transcription Factor Assay is a non-radioactive, sensitive method for detecting CREB DNA binding activity. CREB contained in a nuclear extract or whole cell lysate binds specifically to the DNA cAMP response element immobilized to the wells a 96-well plate. The activated CREB transcription factor complex is detected by addition of a specific primary antibody directed against Phospho-Ser133 on CREB. A secondary antibody conjugated to HRP is used to provide a sensitive colorimetric readout at 450 nm.
Delivery information: Kit contains buffer, reagent A, positive control, binding buffer, primary antibody, buffer concentrate, polysorbate 20, competitor dsDNA, HRP conjugate, plate and cover sheet, developing and stop solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty