Would you like to log in again?
His Capture Kit, Cytiva
Supplier: Cytiva
28-9950-56
102985-700EA 1340.23 USD InStock
102985-700
His Capture Kit, Cytiva
Assays Protein Assays
Reagent kit for the capture of histidine-tagged ligands in Biacore systems, providing an alternative to capture on nickel-chelated nitrilotriacetic (NTA) groups.
  • Easy: minimum assay development with no need to identify reagents or verify assay conditions
  • Convenient: optimized protocol and all required solutions for the efficient capture of histidine-tagged proteins included
  • For use with: enables antibody capture of histidine-tagged ligands in Biacore systems
Depending on the nature of the interacting molecules, this approach might give lower non-specific binding and/or higher binding stability. The anti-histidine antibody is a monoclonal antibody (MAb) directed against polyhistidine tags in the C or N terminus of a protein.
Ordering information: The use of these products in Biacore systems requires an amine coupling kit with sensor chip CM5, CM4, CM3, or C1. Not for use with Biacore Flexchip.
Delivery information: It includes anti-histidine antibody, immobilization buffer, and regeneration solution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty