Would you like to log in again?
Edge® 48-128 Industrial Work Glove with Nitrile Coating, Ansell
Supplier: Ansell
Edge®
48128100 48128080 48128070 48128090 48128060 48128110
76196-312CS 844.78 USD InStock
76196-312 76196-322 76196-320 76196-318 76196-316 76196-314
Edge® 48-128 Industrial Work Glove with Nitrile Coating, Ansell
Gloves
A better alternative to light or medium weight knitted cotton gloves due to the nitrile coating that provides much better grip, cushioning, and abrasion resistance.
  • Very light and comfortable
  • High level of dexterity and flexibility
  • Good level of abrasion and oil resistance
  • Dark glove shows dirt contamination less quickly
These industrial work gloves combine a seamless knitted dark grey polyester liner with an elasticized knit wrist dipped into a black straight nitrile coating. A top quality grip along with good abrasion resistance make this glove the practical choice for light applications where oil is present.
Certifications: CE marked. EN 388:2003 - 4121. EN 388:2016 - 4121A. REACH compliant.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty