Would you like to log in again?
Septa for 96-Well Plates, Cytiva
Supplier: Cytiva
29192561
103568-388EA 228.32 USD InStock
103568-388
Septa for 96-Well Plates, Cytiva
Seals Microplate Seals
Septa used to cover microplates in experiments where the injection needles enter each well more than once. For use with Biacore 8k, Biacore T200 and Biacore S200.
  • Foil made of OPP (oriented polypropylene)
  • Rubber mat made of styrene ethylene butylene styrene block copolymer
  • Adhesive: Polyester film (PET) with acrylic adhesive
  • For use with 96-well or 96-deep-well microplates
  • Compatible with polystyrene and polypropylene
Each well is resealed and the needles are wiped off when elevated through the septa, ensuring high-quality data.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty