Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TBK1 Mouse Monoclonal Antibody [clone: 4E6]

Supplier: Cayman Chemical
25924-100
76304-456EA 426.97 USD InStock
76304-456
Anti-TBK1 Mouse Monoclonal Antibody [clone: 4E6]
Antibodies
Type: Primary
Antigen: TBK1
Clonality: Monoclonal
Clone: 4E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty