Would you like to log in again?
Cut Resistant Sleeve, Portwest
Supplier: Portwest
A690GRR
76103-426EA 10.71 USD InStock
76103-426
Cut Resistant Sleeve, Portwest
Workwear
Cut Resistant Sleeve is specially made for superior protection.
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. AS/NZS 2161 .4. ANSI/ISEA 105 - 2016 CUT Level (A4). EN 388:2003 - 4.5.4.X. EN 388:2016 - 4.X.4.X.D. EN 407 X.1.X.X.X.X. EN 420. CE food safe.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty