Would you like to log in again?
Double Palm Rigger Gloves, Portwest
Supplier: Portwest
A230GRRXL
76103-530EA 6.98 USD InStock
76103-530
Double Palm Rigger Gloves, Portwest
Gloves
Providing reinforced protection over the palm and forefinger, this glove is specially made for use in construction, landscaping, agriculture and forestry.
  • Superior double palm rigger
  • Knuckle back protection
  • Cotton backing for breathability
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. AS/NZS 2161 .4. ANSI/ISEA 105 - 2016 ABRASION Level (2). EN 420. EN 388:2003 - 4.2.2.3. EN 407 4.3.X.X.X.X.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty