Would you like to log in again?
Welders Gauntlet, Portwest
Supplier: Portwest
A500RERXL
76103-624EA 12.5 USD InStock
76103-624
Welders Gauntlet, Portwest
Gloves
The tough 14" split leather gauntlet providing protection against a range of heat related tasks. Ideal for welding and metal handling due to the premium quality materials used.
  • EN12477 Type A welding protection
  • Fully lined with soft and comfortable cotton
  • 14 inch cow split leather gauntlet
  • Maximum EN407 burn behaviour resistance
  • Durable leather
Ideal for welding and metal handling.
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. AS/NZS 2161 .4. ANSI/ISEA 105 - 2016 ABRASION Level (3). EN420. EN388:2003 - 3.2.3.3. EN407 4.2.3.X.3.X. heat and fire protection. EN12477 Type A.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty