Would you like to log in again?
Sensor Chip Protein L, Cytiva
Supplier: Cytiva
29205137 29205138
103568-404EA 498.38 USD InStock
103568-404 103568-386
Sensor Chip Protein L, Cytiva
Electrodes Other Electrodes
A Biacore Extend sensor chip for site-directed antibody capture. Suitable for study of a wide range of antibody fragments including Fabs and dAbs.
  • Used for oriented capture of antibody fragments
  • Ready-to-use, eliminating the need to develop immobilization and regeneration conditions, saving time and effort
  • Captures a wide range of antibody fragments and domain antibodies
Carboxymethylated dextran matrix pre-immobilized with a recombinant Protein L for antibodies and antibody fragments containing kappa light chain subtypes (1, 3 and 4) without interfering with its antigen-binding site.
Protein L is suitable for the capture of a wider range of antibody classes than both Protein A and Protein G, including IgG, IgM, IgA, IgE, and IgD. This does not bind bovine immunoglobulins, which often contaminate serum supplements, and is therefore especially useful for quantifying antibodies and antibody fragments from serum based culture.
'S series' sensor chips are for use with Biacore 8K, Biacore S200, Biacore T200, Biacore T100, and Biacore 4000 SPR systems.
Sensor chips not designated with 'S series' are designed for Biacore X100, Biacore 3000 and Biacore C SPR systems.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty