Would you like to log in again?
Whatman™ Application-Specific Filter Papers, Grade 0048, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10348903 10348903
10035-660PK 348.86 USD InStock
10035-660 CA11013-410
Whatman™ Application-Specific Filter Papers, Grade 0048, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Filter mat made from a mixture of cellulose and polyester. This mat is used for optically testing baby food (artificial milk) for textile fibers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty