Would you like to log in again?
Whatman™ Application-Specific Filter Papers, Grade 0790 ½, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10301645 10301647 10301645 10301647
10035-606PK 294.56 USD InStock
CA11012-590 10035-606 CA11012-610 10034-632
Whatman™ Application-Specific Filter Papers, Grade 0790 ½, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Acid washed cellulose filter paper with ash content of approximately 0,01%, low magnesium, for the determination of trace elements (Mg, Mn, Co, Cu, Mo, B).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty