Would you like to log in again?
Whatman™ Application-Specific Filter Papers, Grade 3459, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Whatman™
10316619 10316619
10034-742PK 303.76 USD InStock
CA11012-644 10034-742
Whatman™ Application-Specific Filter Papers, Grade 3459, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Creped cellulose filter papers with good retentivity at a relatively high filtration speed. They are used for the clarifying filtration of dried beet pulp extracts or beet juice after the addition of lead acetate for subsequent polarimetric sugar determination. They are specifically designed for the Venema unit (lead acetate method).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty