Would you like to log in again?
Whatman™ Application-Specific Filter Papers, Grade 287 ½, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10310245 10310247 10310245 10310247
10035-612PK 86.54 USD InStock
CA11012-586 10035-612 CA11012-588 10035-614
Whatman™ Application-Specific Filter Papers, Grade 287 ½, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Kieselguhr paper with a medium to slow flow rate. Additional adsorption effect, e.g., for the separation of very fine semi-colloidal turbidity, for clarifying milk serum, starch solutions, soil suspensions or sugar-containing solutions prior to polarimetry or refractometry.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty