Would you like to log in again?
Whatman™ Application-Specific Filter Papers, Grade 512 ½, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Whatman™
10310643 10310647 10310643 10310645 10310645 10310647
10035-616PK 91.9 USD InStock
10035-616 10035-618 CA11012-640 CA11012-874 CA11012-576 10035-620
Whatman™ Application-Specific Filter Papers, Grade 512 ½, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Low phosphate cellulose papers approximately 1,5 ppm phosphate, for the filtration of calcium lactate extracts from soil samples for the determination of K and P according to Egnér, Riehm and Lederle.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty