Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water Premeasured, One Solution st™ Vesphene III for injection (WFI)

Supplier: STERIS CORP
Synonyms: Dihydrogen oxide

1S15CX
76303-148CS 394.37 USD InStock
76303-148
Water Premeasured, One Solution st™ Vesphene III for injection (WFI)
Water
The One Solution st Vesphene III System Pre-measured WFI combines a sterile unit dose pouch with a gallon of USP Water For Injection (WFI).

  • Efficacy - Phenolic active ingredients provide broad spectrum antimicrobial efficacy
  • Convenience - Gamma irradiated and double bagged for easy introduction into your aseptic areas
  • Flexibility - Ideal for use in a contamination control rotation
  • Reliability - Complete lot traceability
  • Assurance - Each lot documented by Certificate of Analysis

The family of One Solution st Water For Injection products meets United States Pharmacopeia Water For Injection (WFI) specifications and is intended to be used to dilute surface disinfectants and cleaners.

Sterile filtered and filled in a controlled environment, each pouch of disinfectant attached to a gallon of USP water for injection and double bagged and gamma irradiated through a validated process for easy introduction into aseptic areas.

The use-dilution can be applied manually or sprayed.


Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty