Would you like to log in again?
LEYBOLD® Extraction of Copper from Copper Oxide
Supplier: Leybold
C1.5.1.1
470311-238EA 1477.89 USD InStock
470311-238
LEYBOLD® Extraction of Copper from Copper Oxide
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
LEYBOLD® Chemistry Experiments.
  • Metals and their properties
  • Extraction of metals
  • Non-metals and their properties
Charcoal is the reducing agent to reduce tenorite (CuO) to elementary copper. When heated, the mixture of tenorite and pulverized charcoal produces red copper and carbon dioxide. A saturated calcium hydroxide solution is used to verify the presence of the carbon dioxide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty