Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ellipso-Spoon® Spatula, Chemglass
Supplier: Chemglass
CG-1987-04
89028-940EA 221.69 USD InStock
89028-940
Ellipso-Spoon® Spatula, Chemglass
Spatulas
The specialty curved shape permits contact with all corners of bottles, flasks, and containers.
  • Made of polished stainless steel
  • Handle diameter is 8 mm
Crystals, powders, solid substances and thick liquids are easily retrieved and do not fall back into the container. The sampler has a spoon and spatula.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty