Would you like to log in again?
Esteem™ Comfort Nitrile Exam Gloves, Cardinal
Supplier: Cardinal
88ES04L
76311-752BX 78.86 USD InStock
76311-752
Esteem™ Comfort Nitrile Exam Gloves, Cardinal
Gloves
Esteem™ Comfort are the newest generation of Esteem™ stretchy nitrile exam gloves.
  • 30% higher minimum elongation than the minimum ASTM standard for a nitrile exam glove
  • Reduce cuff tears during donning
  • Reduces the risk of developing a Type IV hypersensitivity to chemical accelerants
  • Tested against chemotherapy drugs per ASTM D6978
  • Tested against 18 chemicals
  • Textured fingertips for enhanced grip
These gloves offer the perfect combination of improved elongation with outstanding protection from chemotherapy drugs, and an FDA-approved low dermatitis potential claim.
Certifications: Gloves meet ASTM Standards for D6319, D6978, and F739.
Caution: Gloves used for protection against chemotherapy drug and laboratory chemical exposure should be selected specifically for the type of chemicals being used. Review Safety Data Sheets (SDS) for the chemicals/drugs being used to determine required level of protection.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty