Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water ACS, meets ASTM Type II specifications for preparative liquid chromatography, general laboratory use

Supplier: Honeywell Research Chemicals
Synonyms: Dihydrogen oxide

LP366-5G LP366-4P
BJLP366-5GEA 206.5 USD InStock
BJLP366-5G BJLP366-4P
Water ACS, meets ASTM Type II specifications for preparative liquid chromatography, general laboratory use
Water
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.9971 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now
Specification Test Results
Chloride (Cl) 5 µg/L max.
Electrical Conductance (25ºC) 1 µS/cm max.
Electrical Resisitivity (25ºC) 1 MOhm/cm min.
Sodium (Na) 5 µg/L max.
Total Organic Carbon 50 µg/L max.
Total Silica (SiO2) 3 µg/L max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty