Would you like to log in again?
3B Scientific® Lecture Hall Ear
Supplier: AMERICAN 3B SCIENTIFIC SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
3B Scientific®
1001266
470311-834EA 12225 USD InStock
470311-834
3B Scientific® Lecture Hall Ear
Models Human Biology Models
Is this possibly the worlds largest ear?
  • 15x life size
  • Includes base
  • Three year manufacturers warranty
  • Includes 3B Smart Anatomy
At 15 times life-size, this 3-part human ear is suitable for museums and special collections as well as large lecture halls and conferences. The giant ear represents the outer, middle and inner ear. The auditory ossicles and the labyrinth with cochlea and vestibulocochlear nerve can be removed from the ear and studied in detail.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty