Would you like to log in again?
Chelex Beads, G-Biosciences
Supplier: G-Biosciences
BTNM-0024
76287-000EA 63.09 USD InStock
76287-000
Chelex Beads, G-Biosciences
Beads Plastic Beads
Chelex beads are made of a styrene divinylbenzene copolymer containing paired iminodiacetate ions, which act as chelating groups in binding polyvalent metal ions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty