Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
PharmaTainer™ Carboys, PET, BIOFLUID FOCUS
Supplier: SANISURE MS
120000-01
MSPP-120000-01CS 307.31 USD InStock
MSPP-120000-01
PharmaTainer™ Carboys, PET, BIOFLUID FOCUS
Bottles Carboys
These bottles are designed for single-use, bioprocess applications such as collection, storage and transport of bulk vaccines, biopharmaceuticals, other biologics, culture media and more.
  • Ultra-clean, molded and capped in ISO Class 5 environment
  • Eliminates cleaning and concerns about carryover
  • Graduations and traceability information printed on bottle
Precision-formed necks and double-seal HDPE closure system provides reliable, repeatable sealing.
Certifications: Certified below USP 788 standard for particulate and pass USP-VI, in vitro cytotoxicity and endotoxin tests.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty