Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzoic acid 99.5-100.5% (by acidimetry on anhydrous substance), powder, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, E210, SAFC®

Supplier: MilliporeSigma
Danger

1.00130.9025
EM1.00130.9025EA 1031 USD InStock
EM1.00130.9025
Benzoic acid 99.5-100.5% (by acidimetry on anhydrous substance), powder, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, E210, SAFC®
Benzoic acid
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental impurities). Class 1 to 3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated.

Conforms to the purity criteria on food additives according to the current European Commission Regulation.


Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 121…123 °C
Density: 1.316 g/cm³ (20 °C)
Flash Pt: 121 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002398
CAS Number: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/11/748111.jpg
Order Now
Specification Test Results
Assay (Acidimetric) 99.5 - 100.5%
Assay (Acidimetric, Calculated on Anhydrous Substance) 99.5 - 100.5%
Identity Conforms
Appearance of Solution (5%; Ethanol 96%) Passes Test
pH-Value (Saturated Solution) 2.5 - 3.5
Solidification Point (Lower Value) ≥121 °C
Solidification Point (Upper Value) ≤123 °C
Sulfate (SO₄) ≤0.02%
As (Arsenic) ≤1 ppm
Cd (Cadmium) ≤10 ppm
Cr (Chromium) ≤2.5 ppm
Cu (Copper) ≤10 ppm
Hg (Mercury) ≤1 ppm
Mn (Manganese) ≤25 ppm
Ni (Nickel) ≤2 ppm
Pb (Lead) ≤2 ppm
Zn (Zinc) ≤10 ppm
Halogenated Compounds (as Cl) ≤0.01%
Toluene (HS-GC) ≤890 ppm
Other Residual Solvents (ICH Q3C) Excluded by the Production Process
Oxidizable Matter (Ph. Eur.) Passes Test
Oxidizable Matter (USP) Passes Test
Readily Carbonisable Substance Passes Test
Water ≤0.5%
Sulfated Ash ≤0.05%
Corresponds to Ph. Eur., BP, USP, E210

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty