Would you like to log in again?
Screw Cap Micro Tubes with Printed Labels, Sarstedt
Supplier: Sarstedt
72.694.406 72.703.600
MSPP-72694406PK 389.69 USD InStock
MSPP-72694406 MSPP-72703600
Screw Cap Micro Tubes with Printed Labels, Sarstedt
Tubes Microtubes
Micro tubes are with swith skirted base, knurls, assembled cap, writing space and graduation.
Tubes and caps are manufactured from polypropylene and Ethylene propylene diene rubber (EPDM) O-ring.
Certifications: CE certified, PCR Performance Tested (free of DNase/RNase, DNA and PCR inhibitors).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty