Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Labetalol hydrochloride USP reference standard

Supplier: USP
Danger
Synonyms: Normodyne2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzamide hydrochlorideTrandate

1356654
101095-290EA 420 USD InStock
101095-290
Labetalol hydrochloride USP reference standard
Labetalol hydrochloride


Formula: C₁₉H₂₄N₂O₃·HCl
MW: 364.87 g/mol
Melting Pt: 180 °C (dec.)
MDL Number: MFCD00057663
CAS Number: 32780-64-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty