Would you like to log in again?
HCCA Matrix, Bruker
Supplier: Bruker
8255344
MSPP-8255344EA 530.77 USD InStock
MSPP-8255344
HCCA Matrix, Bruker
Protein Purification Protein Purification Reagents
Pre-portioned HCCA matrix provides easy and convenient preparation of HCCA matrix solutions.
  • Stable and easy to handle for sample preparation
HCCA has been developed especially for Bruker MALDI instruments and purified to ensure accurate results and maintain system cleanliness. This reduces frequency of preventive maintenance cleans required.
Once dried on the sample, the small highly homogeneous crystals enable accurate identification of microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty