Would you like to log in again?
HF Acid Eater Spill Kit, Cole Parmer
Supplier: Cole Parmer
86295-04
MSPP-EW8629504EA 325.51 USD InStock
MSPP-EW8629504
HF Acid Eater Spill Kit, Cole Parmer
Spill Control Materials
HF acid eater completely destroys the hydrofluoric acid polymer. This neutralizer has a 1:1 neutralization ratio as apposed to the 12:1 ratio of caustic soda.
  • Non-hazardous and non-corrosive
  • Neutralizes vapors and reduces odors
  • Excellent for a variety of uses and applications
pH indicator additive eliminates the need for additional pH strips taking out the guess work. Can be applied on horizontal, vertical, and perforated surfaces.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty