Would you like to log in again?
Acid Sphingomyelinase Colorimetric Assay Kit II, BioVision
Supplier: BioVision
AB252889-100TEST
76312-788EA 697.13 USD InStock
76312-788
Acid Sphingomyelinase Colorimetric Assay Kit II, BioVision
Assays Enzyme Assays
Complete kit that allows the measurement of acid sphingomyelinase in biological samples.
  • Complete set of reagents for the detection of acid sphingomyelinase in biological samples
  • Simple and specific (detect acid SMase activity only)
Sphingomyelinase (SMase) cleaves sphingomyelin to produce phosphorylcholine and ceramide. The activation of SMase leads to increased production of ceramide, which acts as a lipid second messenger. There are five distinct types of SMases. Deficiency in acid SMase (ASMase) leads to niemann-pick disease type A and B.
BioVision’s acid sphingomyelinase activity assay kit provides a simple and sensitive method for measuring ASMase enzymatic activity using colorimetry (OD 570 nm). In this assay, ASMase converts its substrate, sphingomyelin to phosphorylcholine and ceramide at pH 5.0; subsequently, phosphorylcholine is utilized in a series of reactions culminating in color formation from a highly specific probe. This high-throughput adaptable assay kit can detect ASMase activity as low as three mU/ml in a variety of samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty