Would you like to log in again?
Phosphoglycerate Kinase Activity Assay Kit (Colorimetric), BioVision, Inc.
Supplier: BioVision
AB252890-100TEST
76312-844EA 957.62 USD InStock
76312-844
Phosphoglycerate Kinase Activity Assay Kit (Colorimetric), BioVision, Inc.
Assays Enzyme Assays
First kit in the market to measure PGK activity in biological samples.
  • Sensitive (~50 mU)
  • Fast and convenient
  • Protocol takes ~60 minutes to be completed
Phosphoglycerate kinase (PGK1) (EC 2.7.2.3) participates in two of the most important biochemical pathways in living organisms: Glycolysis and gluconeogenesis. PGK is responsible for the catalysis of the reversible reaction of 1,3-bisphosphoglycerate (1,3-BPG) and ADP to 3-phosphoglycerate (3-PG) and ATP. In humans, PGK1 and PGK2, two PGK isozymes, have been characterized. While PGK1 is X-Chromosome-linked variant and expressed in all cells, PGK2 is autosome-linked variant and uniquely expressed in spermatogenic cells.
Recent studies have shown deficiency of phosphoglycerate kinase could lead to chronic hemolytic anemia, mental disorders and myopathy in humans, whereas PGK overexpression promotes gastric cancer cell invasiveness. Therefore, accurate measurement of phosphoglycerate kinase activity is useful for mechanistic and possible diagnostic studies.
BioVision’s PGK activity assay kit provides a quick and easy way for monitoring PGK activity in various samples. In the first step of this enzymatic assay, PGK converts 3-phosphoglycerate and ATP to 1,3-bisphosphoglycerate and ADP. The nascent intermediate is detected via a series of enzymatic reactions that lead the oxidation of NADH to NAD+, which can be easily detected.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty