Would you like to log in again?
PicoProbe™ Glycerol-3-Phosphate Assay Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB252891-100TEST
76312-846EA 957.6 USD InStock
76312-846
PicoProbe™ Glycerol-3-Phosphate Assay Kit (Fluorometric), BioVision, Inc.
Assays Enzyme Assays
Highly sensitive method, includes paraoxonase positive control.
  • Simple, sensitive and rapid
  • This assay kit can detect G3P less than 20 pmol/well and can be used for a variety of sample types
Glycerol-3-phosphate (G3P) is an important intermediate for all living organisms. Glycerol-3-phosphate is produced either by glycerol via glycerol kinase or by glycerol-3-phosphate dehydrogenase from dihydroxyacetone phosphate. In response to cellular signals, glycerol-3-phosphate can be utilized in multiple pathways: it can be further converted into glyceraldehyde-3-phosphate and enter glycolysis, rapidly generate NAD+ in brain or muscle tissues through the G3P shuttle, or enter the lipid biosynthetic pathway. Recent studies have found that glycerol-3-phosphate is a novel regulator and plays a fundamental defense role in plant pathogenesis.
BioVision’s PicoProbe™ glycerol-3- phosphate assay kit is a sensitive, fast and easy-to-use method of measuring G3P concentration in various biological samples. In this assay, G3P is converted into an intermediate by the G3P enzyme mix. The G3P developer then utilizes the intermediate to generate a fluorometric signal with PicoProbe™, which can be used to calculate the concentration of G3P in the sample. This assay kit can detect G3P as little as 20 pmol/well and can be used for a variety of sample types.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty