Would you like to log in again?
Arginine Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB252892-100TEST
76312-854EA 867.24 USD InStock
76312-854
Arginine Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
Highly sensitive assay for the measurement of arginine in serum.
  • Simple procedure; takes ~120 minutes
  • Kit contains the necessary reagents for accurate measurement of arginine
  • Fast and convenient
L-arginine (Arg) is a proteogenic, semi-essential amino acid: Healthy humans can synthesize L-arginine using L-glutamine as a building block. However, premature infants are unable to produce arg and additional supplementation is required for proper growth and development. Arginine plays pivotal roles in biochemical pathways such as the urea cycle and the biosynthesis of nitric oxide. Arginine and ammonia concentrations are elevated in patients having a mutation in their ARG1 genes. The mutation causes lower arginase activities- a condition that is known as argininemia. Arginine has also been advertised as a supplement due to its role in the synthesis of nitric oxide, which helps in vasodilation processes.
BioVision’s L-arginine assay kit provides a quick, specific and easy method for the measurement of total L-arginine concentrations in a wide variety of samples. In our enzyme-based assay, L-arginine is converted into a series of intermediates, which will further react with a probe producing a stable fluorescence signal (Ex/Em = 535/587 nm). The kit is simple to perform, specific, high-throughput adaptable and sensitive- it can detect as low as 100 pmol/well of L-arginine in human biofluids and other biological samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty