Would you like to log in again?
PicoProbe™ Glutamate Assay Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB252893-100TEST
76312-856EA 957.35 USD InStock
76312-856
PicoProbe™ Glutamate Assay Kit (Fluorometric), BioVision, Inc.
Assays Enzyme Assays
Highly sensitive; validated.
  • Simple procedure; takes ~60 minutes
  • Fast and convenient
  • The kit provides an accurate assay for measuring inositol in various samples
Glutamate (glutamic acid, glu) is one of the most abundant proteogenic amino acids in humans. It is a key molecule in cellular metabolism. For example, glutamate plays an important role both in the metabolism of some amino acids and disposal of excess nitrogen in the form of urea. In addition, glutamate is the main excitatory neurotransmitter in mammalian nervous system and involved in learning, cognitive and memory processes. Glutamate signaling activates receptors which have been implicated in mental disorders (schizophrenia), neurodevelopment disorders (autism) and neurodegenerative diseases like alzheimer’s and parkinson’s disease.
BioVision’s PicoProbe™ glutamate assay kit (fluorometric) allows for quantification of glutamate in biological fluids and tissues. The assay is based on an enzymatic reaction in which a fluorogenic probe is reduced producing a stable signal. The reduced fluorophore produces a strong signal (Ex/Em= 535/587 nm), which is directly proportional to the amount of glutamate in samples. The assay is simple, reproducible, and can specifically detect as low as 5 pmol of glutamate in a 100 µl reaction.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty