Would you like to log in again?
DNA Quantification Assay Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB252898-2000TEST
76312-868EA 957.6 USD InStock
76312-868
DNA Quantification Assay Kit (Fluorometric), BioVision, Inc.
Assays Enzyme Assays
Sensitive, specific assay. Protein, RNA and ssDNA do not react with the probe.
  • Simple, one-step procedure
  • Results in 30 mins or less
  • Sensitive and specific
  • Does not detect protein, RNA or ssDNA
DNA quantification is essential for a variety of applications, such as real-time PCR (rtPCR), qPCR, Next Generation Sequencing (NGS) and more. The measurement of DNA using analytical instruments (i.e. 'nanodrop') is often limited by the presence/contamination of other macromolecules like protein and RNAs found in biological samples. However, the development of highly specific probes that bind to DNA allows the measurement of this macromolecule using a fluorimeter. The biggest advantages of this approach is protein, RNA and single-stranded DNA (ssDNA) are not detected.
BioVision’s DNA quantification assay kit provides a quick, specific, and easy method for the measurement of DNA concentrations in a wide variety of samples. In this assay, DNA specifically reacts with a probe producing a stable fluorescence signal (Ex/Em = 492/528 nm). The kit is simple to perform, specific, sensitive and high-throughput adaptable with a wide detection range (0.4 to 150 ng DNA) and a limit of detection of 4 pg/µl dsDNA in samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty