Would you like to log in again?
Ornithine Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB252903-100TEST
76312-876EA 957.6 USD InStock
76312-876
Ornithine Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
Kit that measures ornithine in biological samples.
  • Kit contains the necessary reagents for accurate measurement of ornithine
  • Specific and sensitive
  • Simple procedure, takes ~120 minutes, fast and convenient
Ornithine is a non-standard, non-proteogenic amino acid. Ornithine is converted to putrescine via ornithine decarboxylase, which is the first step in the synthesis of polyamines. Additionally, it is one of the main metabolites that participate in the urea cycle in order to excrete excess nitrogen in the form of urea. In cells, arginase hydrolyzes arginine producing ornithine and urea (cytosol). Then, ornithine enters the mitochondria, reacts with carbamoyl phosphate forming citrulline. Ornithine has therapeutics uses: It is administered to patients suffering brain complications caused by hepatic encephalopathy. Additionally, early research has suggested ornithine may improve performance in some professional athletes.
BioVision’s ornithine assay kit provides a rapid, specific, and easy method for the measurement of total ornithine concentrations in a wide variety of biological samples. In this enzyme-based assay, ornithine is converted into a series of intermediates, which will further react with a probe producing a stable fluorometric signal (Ex/Em = 535/587 nm). The kit is simple to use, sensitive and high-throughput adaptable and can detect as low as 50 pmol/well of ornithine in biological samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty