Would you like to log in again?
Protease Assay Kit, EMD Millipore
Supplier: MilliporeSigma
Calbiochem®
539125-1KIT
EM539125-1KITEA 1100.65 USD InStock
EM539125-1KIT
Protease Assay Kit, EMD Millipore
Assays Protein Assays
This kit suitable for quantitative array of a wide variety of proteases in biological samples, utilizing FTC-casein as a substrate.
  • Protect from light
  • SUitable for colorimetric detection method
Used to detect the enzyme activities of chymotrypsin, elastase, plasminogen, PRONASE® Protease, subtilisin, thermolysin and trypsin.
Delivery information: Kit contains fluorescent-casein substrate, incubation buffer, assay buffer trypsin standard, and a user protocol.
Caution: FTC-casein, protease positive control, incubation buffer should be stored at –20 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty