Would you like to log in again?
Human Type AB Serum, Heat Inactivated, Valley Biomedical
Supplier: Valley Biomedical
HUMANABSRMP-HI-1
MSPP-HP1022HIEA 223.08 USD InStock
MSPP-HP1022HI
Human Type AB Serum, Heat Inactivated, Valley Biomedical
Sera (Serum)
Human Type AB Serum is used as a growth supplement for cell culture media and in other research applications.
  • Collected from human AB plasma which is collected from healthy male donors at FDA-licensed facility
Tested and found negative for all required viral markers such as HBsAg, HIV-1, HIV-2, HCV, HIV-1Ag or HIV-1 NAT, ALT and syphilis.
Heat inactivation protocol is to bring the temperature of the material to 56 °C and maintain this for 30 minutes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty