Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hemoglobin A1c (HbA1c) (Human) ELISA Kit, BioVision

Supplier: BioVision
AB289836-96TEST
76313-358EA 1312.37 USD InStock
76313-358
Hemoglobin A1c (HbA1c) (Human) ELISA Kit, BioVision
Assays ELISAs
A Sandwich ELISA kit for the quantitative measurement of Hemoglobin A1c in biological fluids.

  • Easy, convenient and time-saving method to measure the level of HbA1c in serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • Detection Range: 3125 to 200 ng/ml
  • Sensitivity: <1875 ng/ml
  • Assay Precision: Intra-Assay: CV <8%; Inter-Assay: CV <10%
  • No significant cross-reactivity or interference was observed

Glycated hemoglobin (HbA1c) is a form of hemoglobin that is measured primarily to identify average plasma glucose concentration. HbA1c is a measure of the beta-N-1-deoxy fructosyl component of hemoglobin. Glycated hemoglobin testing is recommended for both checking the blood sugar control in people who might be pre-diabetic and monitoring blood sugar control in patients with more elevated levels, termed diabetes mellitus. It provides far more revealing information on glycemic behavior than a fasting blood sugar value. BioVision’s HbA1c ELISA kit is a sandwich ELISA assay for the quantitative measurement of HbA1c in human serum, plasma and cell culture supernatants. The density of color is proportional to the amount of HbA1c captured from the samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty