Would you like to log in again?
NMR Tube Liners, New Era™
Supplier: New Era
NE35010 NE3505
100165-890EA 39.77 USD InStock
100165-890 100165-926
NMR Tube Liners, New Era™
Tubes NMR Tubes
An inexpensive way to handle corrosive samples that may react with the glass sample tube.
Thin wall, round bottom liners are supplied with a PTFE plug closure. For use in standard thin wall 5 and 10 mm sample tubes. Results may vary and some higher order shimming may be required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty