Would you like to log in again?
Sample Tube Caps, PTFE, New Era™
Supplier: New Era
NE312510
100165-906PK 70.69 USD InStock
100165-906
Sample Tube Caps, PTFE, New Era™
Closures Test/Sample Tube Closures
Highly inert 5 mm PTFE sample tube caps.
  • Precision machined cylindrical design maintains good spinning quality
  • Snug push fit ensures solvent retention
  • Circular external groove aids in removal
The easiest way to remove the cap is to grasp under the cap with a finger nail and pull straight up and off. Caps should be cleaned before reuse.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty