Would you like to log in again?
NOS2 / iNOS (Mouse) ELISA Kit, BioVision
Supplier: BioVision
AB285318-96TEST
76313-428EA 1516.4 USD InStock
76313-428
NOS2 / iNOS (Mouse) ELISA Kit, BioVision
Assays ELISAs
A Sandwich ELISA kit for the quantitative measurement of NOS2 in biological fluids.
  • Sensitivity: <0.188 ng/ml, Detection method- Absorbance (450 nm)
  • Detection Range: 0.312 to 20 ng/ml , Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%, Application- quantitative measurement of NOS2 in serum, plasma, tissue homogenates and other biological fluids
  • No significant cross-reactivity or interference was observed,Species reactivity- mouse
Nitric oxide synthases (NOS) are a family of enzymes that catalyze the production of nitric oxide (NO) from L-arginine. Nitric oxide (NO) plays an important role in neurotransmission, vascular regulation, immune response and apoptosis. In presence of NADPH, FAD, FMN, (6R)-5,6,7,8-tetrahydrobiopterin, calmodulin and heme, NOS catalyzes a five-electron oxidation of the guanidino nitrogen of L-arginine with molecular oxygen to generate NO and L-citrulline. There are three isoforms of NOS: endothelial (eNOS), neuronal (nNOS), and inducible (iNOS). iNOS is expressed de novo under oxidative stress conditions and produces large amounts of NO as a part of body’s defense mechanism. BioVision’s NOS2 ELISA kit is a sandwich ELISA assay for the quantitative measurement of NOS2 in mouse serum, plasma and cell culture supernatants. The density of color is proportional to the amount of NOS2 captured from the samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty