Would you like to log in again?
Oakton® Gel Filled pH Electrode, Double-Junction, BNC Cable, Cole-Parmer
Supplier: Cole Parmer
35641-51
76221-300EA 110.39 USD InStock
76221-300
Oakton® Gel Filled pH Electrode, Double-Junction, BNC Cable, Cole-Parmer
Electrodes pH Electrodes
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty