Would you like to log in again?
Snap Caps, Sun Sri
Supplier: SUN SRi
501 312 501 316 501 318 501 313 501 317
14232-490PK 56.28 USD InStock
14232-490 14232-496 14234-668 14234-864 14234-786
Snap Caps, Sun Sri
Closures Test/Sample Tube Closures
These snap caps eliminate crimping and decrimping of closures with easy to apply snap caps and also it provides a secure seal, minimizing evaporation even with volatile samples.
  • Septa pre-assembled
  • Compatible with StepVials and other snap-ring style vials
  • Not compatible with a standard 11 mm aluminum seal finish
The audible click ensures that a secure seal has been formed and the cap is properly applied.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty