Would you like to log in again?
Polypropylene Feeding Needles for Rodents, Instech Laboratories
Supplier: Instech Laboratories
FTP-22-25 FTP-20-38
MSPP-FTP2225PK 976.92 USD InStock
MSPP-FTP2225 MSPP-FTP2038
Polypropylene Feeding Needles for Rodents, Instech Laboratories
Animal Supplies Animal Feeding Needles
The tubes are flexible to reduce trauma, translucent to visualize delivery. Flexible tubes are a recommended animal welfare refinement over rigid metal tubes.
  • Soft tip to minimize trauma
  • Disposable - no cleaning, no risk of residual compound
  • Faster to train new technicians
  • Flexible to reduce the risk of perforation
  • Sterile
The feeding tubes using medical grade materials to minimize the risk of interactions with test compounds. The tube is constructed of polypropylene, the same material used in plastic syringes. The soft elastomer tip is designed to prevent inadvertent placement in the trachea and reduce the risk of perforation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty